KEY458251509 Key Organics / BIONET

2-(3-chlorophenyl)-3-methylquinoline-4-carboxylic acid

  • MDL number MFCD01993648
  • Cas Number : 350997-46-5
  • Molecular Weight : 297.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/66554

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1Y-0958_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart