KEY458251511 Key Organics / BIONET

3H,4H-thieno[3,2-d]pyrimidin-4-one

  • MDL number MFCD03407527
  • Cas Number : 16234-10-9
  • Molecular Weight : 152.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/66568

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1Z-0222_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart