KEY458251514 Key Organics / BIONET

1-(2-chloroethyl)-3,5-dimethyl-1H-pyrazole

  • MDL number MFCD00215355
  • Cas Number : 38460-08-1
  • Molecular Weight : 158.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/66688

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2D-027_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart