KEY458251521 Key Organics / BIONET

(2Z)-3-hydroxy-2-phenylprop-2-enenitrile

  • MDL number MFCD00137446
  • Cas Number : 62914-59-4
  • Molecular Weight : 145.161

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/67070

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2G-025_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart