KEY458251529 Key Organics / BIONET

5-(2-chlorophenyl)-1H-1,2,3,4-tetrazole

  • MDL number MFCD00040950
  • Cas Number : 50907-46-5
  • Molecular Weight : 180.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/67532

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2J-051_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart