KEY458251539 Key Organics / BIONET

methyl 4-benzenesulfonamidobenzoate

  • MDL number MFCD00591212
  • Cas Number : 107920-79-6
  • Molecular Weight : 291.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/68095

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2L-390S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart