KEY458251541 Key Organics / BIONET

2-(naphthalen-1-yloxy)acetic acid

  • MDL number MFCD00003927
  • Cas Number : 2976-75-2
  • Molecular Weight : 202.209

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/68366

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2M-936_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart