KEY458251543 Key Organics / BIONET

(2H-1,3-benzodioxol-5-yl)thiourea

  • MDL number MFCD00041216
  • Cas Number : 65069-55-8
  • Molecular Weight : 196.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/68379

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2N-008_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart