KEY458251547 Key Organics / BIONET

[(E)-[(1-methyl-1H-pyrrol-2-yl)methylidene]amino]thiourea

  • MDL number MFCD01568653
  • Cas Number : 6823-89-8
  • Molecular Weight : 182.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/68545

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/2N-900_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart