KEY458251555 Key Organics / BIONET

4-{[(thiophen-2-yl)methyl]amino}benzoic acid

  • MDL number MFCD00475037
  • Cas Number : 1715-13-5
  • Molecular Weight : 233.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 172 - 174
Iupac Name 4-[(2-thienylmethyl)amino]benzoic acid
Inchi Code 1S/C12H11NO2S/c14-12(15)9-3-5-10(6-4-9)13-8-11-2-1-7-16-11/h1-7,13H,8H2,(H,14,15)
Inchi Key SHTQXYWTNXSOLW-UHFFFAOYSA-N

Description

Melting Point 172 - 174
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/68615

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/2P-081_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart