KEY458251561 Key Organics / BIONET

6-chloro-3-(difluoromethyl)-[1,2,4]triazolo[4,3-b]pyridazine

  • MDL number MFCD02186392
  • Cas Number : 477889-88-6
  • Molecular Weight : 204.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/69028

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/2R-0679_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart