KEY458251565 Key Organics / BIONET

1-(prop-2-en-1-yl)-2-(trifluoromethyl)-1H-1,3-benzodiazole

  • MDL number MFCD02102641
  • Cas Number : 139591-03-0
  • Molecular Weight : 226.202

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/69269

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/2R-1504_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart