KEY458251570 Key Organics / BIONET

4,4,4-trifluoro-1-(2-methoxyphenyl)butane-1,3-dione

  • MDL number MFCD03419768
  • Cas Number : 15191-69-2
  • Molecular Weight : 246.185

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/69553

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/2T-0855_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart