KEY458251571 Key Organics / BIONET

3-(methylsulfanyl)-4H-1,2,4-triazole

  • MDL number MFCD01312671
  • Cas Number : 7411-18-9
  • Molecular Weight : 115.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/69626

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2T-1506_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart