KEY458251574 Key Organics / BIONET

5-oxo-1-[(1R)-1-phenylethyl]pyrrolidine-3-carboxylic acid

  • MDL number MFCD08690908
  • Cas Number : 915302-94-2
  • Molecular Weight : 233.267

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/69714

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/2W-0339_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart