KEY458251581 Key Organics / BIONET

2-chloro-5-[(1H-1,2,4-triazol-1-yl)methyl]pyridine

  • MDL number MFCD04125022
  • Cas Number : 861211-53-2
  • Molecular Weight : 194.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/69928

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/2X-0848_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart