KEY458251583 Key Organics / BIONET

(2E)-3-(dimethylamino)-1-(thiophen-3-yl)prop-2-en-1-one

  • MDL number MFCD00097929
  • Cas Number : 206563-67-9
  • Molecular Weight : 181.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/69980

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/2Y-0809_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart