KEY458251589 Key Organics / BIONET

3-tert-butyl-4-nitro-1H-pyrazole

  • MDL number MFCD00127341
  • Cas Number : 38858-95-6
  • Molecular Weight : 169.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/70093

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3B-019_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart