KEY458251608 Key Organics / BIONET

2-[(4-fluorophenyl)formamido]acetic acid

  • MDL number MFCD00462263
  • Cas Number : 366-79-0
  • Molecular Weight : 197.165

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/71227

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3J-330S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart