KEY458251611 Key Organics / BIONET

3-chloro-[1,2,4]triazolo[4,3-b]pyridazine

  • MDL number MFCD04125061
  • Cas Number : 33050-36-1
  • Molecular Weight : 154.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/71247

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3J-354S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart