KEY458251613 Key Organics / BIONET

ethyl 3-cyano-7-oxo-4H,7H-pyrazolo[1,5-a]pyrimidine-6-carboxylate

  • MDL number MFCD00793850
  • Cas Number : 64689-83-4
  • Molecular Weight : 232.199

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/71385

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3J-930_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart