KEY458251616 Key Organics / BIONET

(2Z)-2-(N-hydroxyimino)-3-oxo-N-phenylbutanamide

  • MDL number MFCD00794549
  • Cas Number : 127745-40-8
  • Molecular Weight : 206.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/71991

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3M-742_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart