KEY458251624 Key Organics / BIONET

(2E)-1-(4-fluorophenyl)-3-(4-methylphenyl)prop-2-en-1-one

  • MDL number MFCD00017965
  • Cas Number : 313236-80-5
  • Molecular Weight : 240.277

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form Off-white to Yellow Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/72072

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/3N-032_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart