KEY458251626 Key Organics / BIONET

6-chloro-4-methylquinolin-2-ol

  • MDL number MFCD01001246
  • Molecular Weight : 193.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/72193

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3N-560S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart