KEY458251627 Key Organics / BIONET

2,6-dichloro-4-methylquinoline

  • MDL number MFCD01316208
  • Cas Number : 90723-71-0
  • Molecular Weight : 212.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2,6-dichloro-4-methylquinoline
Inchi Code 1S/C10H7Cl2N/c1-6-4-10(12)13-9-3-2-7(11)5-8(6)9/h2-5H,1H3
Inchi Key ZNZUDUAJLJNTLP-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/72194

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/3N-561S_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart