KEY458251636 Key Organics / BIONET

6-(morpholine-4-carbonyl)cyclohex-3-ene-1-carboxylic acid

  • MDL number MFCD01339348
  • Cas Number : 478040-26-5
  • Molecular Weight : 239.271

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form White to Off-white Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/72694

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/3R-0280_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart