KEY458251638 Key Organics / BIONET

methyl 2-methanesulfonylacetate

  • MDL number MFCD00051842
  • Cas Number : 62020-09-1
  • Molecular Weight : 152.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 62
Iupac Name methyl (methylsulfonyl)acetate
Inchi Code 1S/C4H8O4S/c1-8-4(5)3-9(2,6)7/h3H2,1-2H3
Inchi Key RQKDASVRZONLNQ-UHFFFAOYSA-N

Description

Melting Point 62
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/72820

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301 + P312 + P330,P302 + P352,P302 + P352 + P312,P304 + P340,P304 + P340 + P311,P305 + P351 + P338,P332 + P313,P337 + P313,P362,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/3R-0841_SDS.pdf
hcodes H302,H315,H319,H332,H335
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart