KEY458251644 Key Organics / BIONET

methyl 4-(morpholin-4-yl)benzoate

  • MDL number MFCD03012127
  • Cas Number : 23676-05-3
  • Molecular Weight : 221.256

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73077

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/3T-0068_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart