KEY458251645 Key Organics / BIONET

(2E)-3-ethoxy-2-(4-methylbenzenesulfonyl)prop-2-enenitrile

  • MDL number MFCD00129579
  • Cas Number : 32253-83-1
  • Molecular Weight : 251.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73078

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/3T-0076_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart