KEY458251651 Key Organics / BIONET

2-(2-oxopropyl)-2,3-dihydro-1H-isoindole-1,3-dione

  • MDL number MFCD00088346
  • Cas Number : 3416-57-7
  • Molecular Weight : 203.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(2-oxopropyl)-1H-isoindole-1,3(2H)-dione
Inchi Code 1S/C11H9NO3/c1-7(13)6-12-10(14)8-4-2-3-5-9(8)11(12)15/h2-5H,6H2,1H3
Inchi Key STMRGLKPBJVVEG-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73520

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/3W-0868_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart