KEY458251665 Key Organics / BIONET

ethyl 2-[(5-chloro-1,3-benzoxazol-2-yl)sulfanyl]acetate

  • MDL number MFCD00127440
  • Cas Number : 84829-54-9
  • Molecular Weight : 271.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73990

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/4B-124_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart