KEY458251677 Key Organics / BIONET

{5-[(4-bromophenyl)sulfanyl]-1-methyl-3-(trifluoromethyl)-1H-pyrazol-4-yl}methanol

  • MDL number MFCD00172639
  • Cas Number : 318239-56-4
  • Molecular Weight : 367.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/74466

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/4G-353S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart