KEY458251679 Key Organics / BIONET

ethyl 2-phenyl-1,3-thiazole-4-carboxylate

  • MDL number MFCD06205115
  • Cas Number : 59937-01-8
  • Molecular Weight : 233.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/74516

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4G-416S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart