KEY458251683 Key Organics / BIONET

(2E)-3-amino-2-cyano-3-[4-(4-fluorophenyl)piperazin-1-yl]-N-methylprop-2-enethioamide

  • MDL number MFCD00243959
  • Cas Number : 338422-39-2
  • Molecular Weight : 319.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/74774

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/4H-909_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart