KEY458251685 Key Organics / BIONET

2-[(2-chlorophenyl)formamido]-3-methylbutanoic acid

  • MDL number MFCD00700123
  • Cas Number : 1396967-55-7
  • Molecular Weight : 255.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/74867

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/4J-305S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart