KEY458251687 Key Organics / BIONET

2-chloro-6-methyl-5-phenylpyridine-3-carbonitrile

  • MDL number MFCD00231559
  • Cas Number : 10176-63-3
  • Molecular Weight : 228.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/74956

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/4J-404S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart