KEY458251689 Key Organics / BIONET

(1-methyl-3-phenyl-5-{[3-(trifluoromethyl)phenyl]sulfanyl}-1H-pyrazol-4-yl)methanol

  • MDL number MFCD00244951
  • Cas Number : 318247-69-7
  • Molecular Weight : 364.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/75211

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/4K-369S_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart