KEY458251696 Key Organics / BIONET

5-iodo-2-(1H-pyrrol-1-yl)pyridine

  • MDL number MFCD01315196
  • Cas Number : 338748-93-9
  • Molecular Weight : 270.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-iodo-2-(1H-pyrrol-1-yl)pyridine
Inchi Code 1S/C9H7IN2/c10-8-3-4-9(11-7-8)12-5-1-2-6-12/h1-7H
Inchi Key MKIBDODBOFHHHO-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/75796

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/4M-955_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart