KEY458251702 Key Organics / BIONET

(Z)-N-{[2-(4-chloro-3-fluorophenoxy)phenyl]methylidene}hydroxylamine

  • MDL number MFCD02082405
  • Cas Number : 451485-70-4
  • Molecular Weight : 265.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/76164

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/4P-024_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart