KEY458251707 Key Organics / BIONET

1-[(2-chlorophenyl)methyl]-5-oxopyrrolidine-2-carboxylic acid

  • MDL number MFCD02082899
  • Cas Number : 87341-46-6
  • Molecular Weight : 253.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/76268

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4P-519S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart