KEY458251713 Key Organics / BIONET

2-(4-nitrophenyl)imidazo[1,2-a]pyridine

  • MDL number MFCD00276341
  • Cas Number : 3323-26-0
  • Molecular Weight : 239.234

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/76895

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4T-0605_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart