KEY458251744 Key Organics / BIONET

ethyl 4-[(4-fluorophenyl)carbamoyl]-5-methyl-1,2-oxazole-3-carboxylate

  • MDL number MFCD00664764
  • Cas Number : 338761-30-1
  • Molecular Weight : 292.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/78367

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/5G-912_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart