KEY458251754 Key Organics / BIONET

ethyl 4,8-dichloroquinoline-3-carboxylate

  • MDL number MFCD00173351
  • Cas Number : 56824-91-0
  • Molecular Weight : 270.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Hygroscopic
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/78622

Safety Information

signalword Warning
pcodes P232, P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/5J-039_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart