KEY458251761 Key Organics / BIONET

(2E)-3-(dimethylamino)-1-(3-nitrophenyl)prop-2-en-1-one

  • MDL number MFCD00024513
  • Cas Number : 1163721-90-1
  • Molecular Weight : 220.228

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/78853

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/5K-030_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart