KEY458251789 Key Organics / BIONET

1H-pyrazole-1-carbothioamide

  • MDL number MFCD00866405
  • Cas Number : 1794-34-9
  • Molecular Weight : 127.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/80670

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/5T-0826_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart