KEY458251791 Key Organics / BIONET

3,5-bis(benzyloxy)benzoic acid

  • MDL number MFCD00191341
  • Cas Number : 28917-43-3
  • Molecular Weight : 334.371

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/80754

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/5W-0042_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart