KEY458251797 Key Organics / BIONET

ethyl 5-(3-nitrophenyl)-1,2-oxazole-3-carboxylate

  • MDL number MFCD03848619
  • Cas Number : 866040-66-6
  • Molecular Weight : 262.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 5-(3-nitrophenyl)-3-isoxazolecarboxylate
Inchi Code 1S/C12H10N2O5/c1-2-18-12(15)10-7-11(19-13-10)8-4-3-5-9(6-8)14(16)17/h3-7H,2H2,1H3
Inchi Key SSOVVOHZUNJEAK-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/80934

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/5W-0869_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart