KEY458251801 Key Organics / BIONET

(5E)-5-{[4-(methylsulfanyl)phenyl]methylidene}-2-sulfanylidene-1,3-thiazolidin-4-one

  • MDL number MFCD00852408
  • Cas Number : 127378-28-3
  • Molecular Weight : 267.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (5E)-5-[4-(methylsulfanyl)benzylidene]-2-sulfanyl-1,3-thiazol-4(5H)-one
Inchi Code 1S/C11H9NOS3/c1-15-8-4-2-7(3-5-8)6-9-10(13)12-11(14)16-9/h2-6H,1H3,(H,12,13,14)/b9-6+
Inchi Key SEIFTQHMDCKSIB-RMKNXTFCSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/80975

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/5X-0220_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart