KEY458251810 Key Organics / BIONET

methyl 2-[(2-chloro-4-nitrophenyl)sulfanyl]acetate

  • MDL number MFCD07021492
  • Cas Number : 139326-35-5
  • Molecular Weight : 261.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/81116

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/5Y-0818_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart