KEY458251816 Key Organics / BIONET

6-chloro-4-oxo-4H-chromene-3-carbaldehyde

  • MDL number MFCD00139138
  • Cas Number : 42248-31-7
  • Molecular Weight : 208.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-chloro-4-oxo-4H-chromene-3-carbaldehyde
Inchi Code 1S/C10H5ClO3/c11-7-1-2-9-8(3-7)10(13)6(4-12)5-14-9/h1-5H
Inchi Key LIGLNCRTMDRUAO-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/81247

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/6C-042_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart